Biphenyl (PCB No. 0) ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Diphenyl, Phenylbenzene, PCB No. 0, 0-PCB

M-Clarity™ quality level = MQ200

8.03090.1000 8.03090.9045
CA8.03090.0100CS 775.86 CAD
CA8.03090.0100 CA8.03090.1000 CA8.03090.9045
Biphenyl (PCB No. 0) ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Biphenyl (PCB No. 0)
Formula: C₁₂H₁₀
MW: 154.21 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 68…70 °C
Flash Pt: 113
Storage Temperature: Ambient
MDL Number: MFCD00003054
CAS Number: 92-52-4
UN: 3077
ADR: 9 III
Merck Index: 13,03346

Order Now

Specification Test Results

Solubility (20°C) inSoluble
Melting Point 68 - 70 °C
Molar Mass 600 kg/m3
Bulk Density 600 kg/m3
Boiling Point 255 °C (1013 hPa)
Vapor Pressure 0.07 hPa (20 °C)
Flash Point 113 °C
Explosion Limit 0.7 - 3.4 %(V)
Ignition Point 570°C
Hill Formula C12H10
Chemical Formula C6H5C6H5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR