Tetrachloro-p-benzoquinone ≥98.0% (by HPLC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Tetrachloroquinone, 2,3,5,6-Tetrachloro-1,4-benzoquinone, 4-Chloranil, p-Chloranil, 2,3,5,6-Tetrachlorobenzo-1,4-quinone, Tetrachloro-1,4-benzoquinone

M-Clarity™ quality level = MQ200

8.02361.0005 8.02361.0250 8.02361.9010
CA8.02361.0005EA 51 CAD
CA8.02361.0005 CA8.02361.0250 CA8.02361.9010
Tetrachloro-p-benzoquinone ≥98.0% (by HPLC) for synthesis, Sigma-Aldrich®
Tetrachloro-p-benzoquinone
Formula: C₆Cl₄O₂
Melting Pt: 290…293 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00001594
CAS Number: 118-75-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20°C) practically inSoluble
Melting Point 290 - 293 °C (sublimed)
Molar Mass 800 kg/m3
Bulk Density 800 kg/m3
Vapor Pressure 1 hPa (71 °C)
pH 3.5 - 4.5 (100 g/l, H2O, 20 °C) (slurry)
Ignition Point 400°C
Synonyms Tetrachloro-p-benzoquinone
Hill Formula C6Cl4O2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR