Ammonium sulfite 35% in water for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.01221.1000
8.01221.2500
8.01221.9025
CA8.01221.1000CS
284
CAD
CA8.01221.1000
CA8.01221.2500
CA8.01221.9025
Ammonium sulfite 35% in water for synthesis, Sigma-Aldrich®
Ammonium sulphite
Formula:
H₈N₂O₃S Melting Pt: 4…8 °C Density: 1.19 g/cm3 (20 °C) Storage Temperature: Ambient |
CAS Number:
10196-04-0 |
Specification Test Results
Solubility | (20°C) Soluble |
Melting Point | 4 - 8 °C |
Boiling Point | 101 °C |
Density | 1.19 g/cm3 (20 °C) |
pH | 7.5 - 7.6 (H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...