Acrylamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Prop-2-enamide

M-Clarity™ quality level = MQ200

8.00830.0100 8.00830.0500 8.00830.5000
EM8.00830.0100CS 1973.97 CAD
EM8.00830.0100 EM8.00830.0500 EM8.00830.5000 CA8.00830.0100 CA8.00830.0500 CA8.00830.5000
Acrylamide for synthesis, Sigma-Aldrich®
Acrylamide (monomer)
Formula: C₃H₅NO
MDL Number: MFCD00008032
CAS Number: 79-06-1
UN: 2074
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 2040 g/l (25°C)
Melting Point 84 °C
Molar Mass 500 kg/m3
Bulk Density 500 kg/m3
Boiling Point 125 °C (33.3 hPa)
Vapor Pressure 0.009 hPa (25 °C)
Density 1.127 g/cm3 (25 °C)
pH 5.0 - 7.0 (50 g/l, H2O, 20 °C)
Ignition Point 424°C
Synonyms Prop-2-enamide
Hill Formula C3H5NO
Chemical Formula CH2CHCONH2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR