Cyclohexane, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: HexahydrobenzeneHexamethylene

M-Clarity™ quality level = MQ300

1.09666.2511 1.09666.4000 1.09666.2511 1.09666.4000
CA1.09666.4000CS 2802.6 CAD
EM1.09666.4000 EM1.09666.2511 CA1.09666.4000 CA1.09666.2511
Cyclohexane, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Cyclohexane
Formula: C₆H₁₂
MW: 84.16 g/mol
Melting Pt: 7 °C
MDL Number: MFCD00003814
CAS Number: 110-82-7
UN: 1145
ADR: 3,II
https://ca.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg

Order Now

Specification Test Results

Solubility 55 mg/l (20°C)
Melting Point 6 °C
Boiling Point 81 °C (1013 hPa)
Vapor Pressure 103 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.4264 (20 °C)
Explosion Limit 1.2 - 8.3 %(V)
Density 0.78 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 260°C
Viscosity Kinematic 1.26 mm2/s (20 °C)
Grade ACS,ISO,Reag. Ph Eur
Synonyms Hexahydrobenzene, Hexamethylene, Naphthene
Hill Formula C6H12

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR