NOVA-TECH™ 56 Non-Woven Light Weight Cleanroom Wipes, High-Tech Conversions
Supplier: High-Tech Conversions
NT56-98 NT56-99 NT56-1212 NT56-1818
10135-758BG 98.13 CAD
10135-758 10135-760 10135-762 10135-764
NOVA-TECH™ 56 Non-Woven Light Weight Cleanroom Wipes, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
NOVA-TECH ™ 56 light weight wipes are an especially effective wipe. They are tough, abrasion resistant, absorbent, and virtually free of particle generation.
  • Lint-free
  • Cleanroom class 100 (ISO Class 5)
  • Light weight
  • Non-woven
  • Abrasion resistant
  • Absorbant
These wipes are comprised of a synergistic blend of virgin polyester and cellulose that is hydroentangled into a uniform, super-tough fabric. These unique features give NOVA-TECH™ 56 light weight wipes a high degree of absorbency and effectiveness during the process of general wiping. Wipes made withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Basis weight 56 g/m²