Spiral Condenser, Reflux, Ace Glass Incorporated
Supplier: Ace Glass
6041-102 6041-104 6041-106 6041-108 6041-110 6041-112
10099-068EA 464.43 CAD
10099-068 10099-070 10099-072 10099-074 10099-076 10099-078
Spiral Condenser, Reflux, Ace Glass Incorporated
Condensers
Spiral reflux condenser with inlet and outlet hose barbs located on the same side at the top of the condenser body for easy hose connections
These condensers are available in multiple lengths, which allow for greater surface area dependant on the applications needs.