1536-Well Non-binding Microplate, Nonsterile, Greiner Bio-One
Supplier: Greiner Bio-One
782900 782904
89137-768CS 2494.42 CAD
89137-768 89137-770
1536-Well Non-binding Microplate, Nonsterile, Greiner Bio-One
Microplates Deep Well Microplates
Greiner Bio-One's non-binding microplate surface is characterized by low protein, DNA and peptide binding properties which improve data quality especially in homogeneous methods such as Fluorescence Polarization Assays (FPA) or Scintillation Proximity Assays (SPA).
  • Nonsterile
  • Available with solid or clear film well bottoms
  • Non-binding surface
  • No potential for degradation or leaching
This non-binding surface is stable under common assay conditions, there is no potential for degradation or leaching and resultant assay interference. Non-bindingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable