Boiling Water Bath, SBB Aqua Plus Series, Grant
Supplier: GRANT USA INC.
SBBAQUA5PLUSUS SBBAQP12US SBBAQP26US
97055-806EA 2644.92 CAD
97055-806 97055-808 97055-810 97055-812
Boiling Water Bath, SBB Aqua Plus Series, Grant
Baths Boiling Baths
Boiling baths are designed for continuous operation with water above 90 °C
  • Adjustable energy regulator provides steady boiling
  • Constant level device maintains liquid level
  • Robust and reliable designs to withstand everyday wear and tear
  • Choice of sizes to suit individual applications
  • Non-drip polycarbonate lid included as standard
The range is ideal for use in education and teaching as well as industrial applications such as equipment sanitisation and sample preparation for immuno assays.
Two over-temperature cut-outs protect users and the workspaceMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable