Gaskets, NW Flange, Ace Glass Incorporated
Supplier: Ace Glass
12192-02 12192-04 12192-06 12192-22 12192-24 12192-26
10099-158EA 338.11 CAD
10099-158 10099-160 10099-162 10099-164 10099-166 10099-168
Gaskets, NW Flange, Ace Glass Incorporated
Seals Gaskets
NW (DN) flange glass to glass sealing gasket featuring a PTFE carrier and o-ring available in Viton® (–26 to 240 °C) or FVMQ flurosilicone (–60 to 230 °C).
Excellent for deep vacuum work or a maximum of 2 bar pressure.