Nunc® Carrier Plate for Cell Culture Inserts, Thermo Scientific
Supplier: Thermo Fisher Scientific
141086 141078 141080 141082 141008 141002 141004 141006
CA89177-072CS 234.49 CAD
CA89177-072 CA89177-074 CA89177-076 CA89177-078 CA89177-080 CA89177-082 CA89177-084 CA89177-086
Nunc® Carrier Plate for Cell Culture Inserts, Thermo Scientific
Microplates Insert Plates and Inserts
Nunc® Carrier Plates are designed for versatility in your research. The system for manipulating multiple inserts can be used for individual compartment cultures or cultures with shared media. Use with up to 12 or 24 inserts at a time.
  • Meets ANSI™ standards
  • Equipped with alphanumeric well identification
  • Polycarbonate membrane is available in three pore sizes for enhanced flexibility in your research
  • Inserts are ideal for for 3-D imaging; translucent membrane becomes transparent when wet
  • Carrier plates come in two formats that correspond with our 12- and 24-well Thermo Scientific™ Nunclon™ Delta-treated dishes
  • Uniquely engineered carrier plate features a support structure that enables variable height
  • Simply rotate the insert to engage one of the three height-adjusting tabs into the carrier plate supports