Sterile Pair Packed Class 100 Nitrile Gloves, USP797/800 Compliant , TechNiGlove
Supplier: Techniglove
STN200P STN201P STN202P STN203P STN204P STN205P
89415-117CS 813.99 CAD
89415-117 89415-119 89415-121 89415-123 89415-125 89415-127
Sterile Pair Packed Class 100 Nitrile Gloves, USP797/800 Compliant , TechNiGlove
Gloves
TechNiGlove pair packed sterile ambidextrous 12" gloves are Technicleaned for Class 100 compatibility and sterilized by gamma irradiation to meet AAMI and ASTM standards.
  • 12" ambidextrous, sterile
  • L/R wallet pair packed
  • TechNiCleaned, USP797/800 approved
  • ASTM D6978 Chemotherapy drug tested
Techniglove products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive on-line batch testing is availableMore Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable