TechNiPak Class 100 Nitrile Cleanroom Gloves, TechNiGlove
Supplier: Techniglove
TN100PFB TN101PFB TN102PFB TN103PFB TN104PFB TN105PFB
89415-066CS 611.01 CAD
89415-066 89415-068 89415-070 89415-072 89415-074 89415-076
TechNiPak Class 100 Nitrile Cleanroom Gloves, TechNiGlove
Gloves
The TechniPak is the world’s only Plastic (PVC) dispenser box of powder-free, 9.5", Class 100 gloves for controlled environments.
  • Plastic (PVC) dispenser box
  • Class 100
  • 9.5" length, powder-free
  • 100% Nitrile
  • Ambidextrous
Techniglove products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive On-line Batch Testing is available for this product.
The clean PVC box reduces particulate from paper boxes and can be wiped clean in wet applications.
Certifications: COC available on request.
Ordering information: Packaged 100 gloves per TechNiPak PVC box, 10 boxes per case.