TechNiTote Class 100 Nitrile Cleanroom Glove, TechNiGlove
Supplier: Techniglove
TNT1200PFB TNT1201PFB TNT1202PFB TNT1203PFB TNT1204PFB TNT1205PFB
89415-054CS 881.95 CAD
89415-054 89415-056 89415-058 89415-060 89415-062 89415-064
TechNiTote Class 100 Nitrile Cleanroom Glove, TechNiGlove
Gloves
The TechniTote is a plastic clamshell dispenser containing powder-free, ambidextrous, 12", Class 100 nitrile gloves for use in applications requiring a clean, portable dispenser.
  • Plastic (PVC) dispenser case
  • Class 100
  • 12"" powder-free
  • 100% Nitrile
  • Ambidextrous
Techniglove products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive On-line Batch Testing is available for this product.
Certifications: COC available on request.
Ordering information: Packaged 250 gloves per TechNiTote, 4 TechNiTotes per case.