TechNiCleaned Pharmaceutical Class 100 Nitrile Cleanroom Gloves, TechNiGlove
Supplier: Techniglove
TN1000W TN1001W TN1002W TN1003W TN1004W TN1005W TN1000B TN1001B TN1002B TN1003B TN1004B TN1005B
89415-030CS 687.98 CAD
89415-030 89415-032 89415-034 89415-036 89415-038 89415-040 89415-042 89415-044 89415-046 89415-048 89415-050 89415-052
TechNiCleaned Pharmaceutical Class 100 Nitrile Cleanroom Gloves, TechNiGlove
Gloves
TechniCleaned Ambidextrous 12" Class 100 powder-free nitrile gloves are designed for Class 100 environments with cost savings in mind.
  • Class 100
  • 100% Nitrile
  • 12" ambidextrous
  • Powder-free
  • For us in semiconductor, disc drive, flat panel, electronic, and pharmaceutical manufacturing
Techniglove products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive On-line Batch Testing is available for this product.
Manufactured in 2 colors: White and Blue. They deliver the same quality, fit, and function with less stringent requirements for particles and extractables than the TN2000 LP series. Suitable for use in semiconductor, disc drive, flat panel, electronic, and pharmaceutical manufacturing.
Certifications: COC available on request.
Ordering information: Packaged 100 gloves per poly-sealed cleanroom bag, 10 bags per case.