Corning® Bar Coded Storage Tubes, Corning
Supplier: Corning
8500 8501 8502 8509 8510 8503
89168-506CS 1248.11 CAD
89168-506 89168-508 89168-510 89168-524 89168-526 89168-512
Corning® Bar Coded Storage Tubes, Corning
Tubes Microtubes
PP. The 1D/2D storage line is an innovative product with both a linear 1D and 2D barcodes etched on each tube.
  • Maximum information, 14×14 dot 2D barcode
  • Barcode stability, laser-etched, not an attached label
  • Compound compatibility, polypropylene construction throughout, making it inert
Order Now
SPECIFICATIONS
Environmentally Preferable