HBIO Masterflex Tubing for Allegro I Peristaltic Pump System, KD Scientific

Supplier: KD Scientific
785618 785619 785620 785621 785622 785623
10117-514BX 366.45 CAD
10117-514 10117-516 10117-518 10117-520 10117-522 10117-524 10117-526 10117-528 10117-532 10117-536 10117-538 10117-540 10117-542 10117-544 10117-552 10117-554 10117-556 10117-558 10117-560 10117-562 10117-564
HBIO Masterflex Tubing for Allegro I Peristaltic Pump System, KD Scientific
Tubing Peristaltic Pump Tubing
Tubing suitable for food applications where FDA compliance is required.

  • Resistant to corrosives, solvents and oils at elevated temperatures
  • Formulation: Thermal set rubber, Viton B (67% fluorine)
  • Application Compatibility: Acids (Excellent), Alkalies (Excellent)
  • Organic solvents: Test First. Viscous fluids (Good), Sterile fluids (Fair)
  • Comes in opaque/black or clear/transparent tubing
  • Non-toxic

KD Scientific recommends Masterflex tubing to achieve the best performance from your Allegro Peristaltic Pump. The tubing used in the Allegro Peristaltic Pump will depend on the pumping application. It is important to select tubing with appropriate chemical resistance for the liquid being pumped.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR