Legato 110 DRS Dual Rate Syringe Pump System, KD Scientific
Supplier: KD Scientific
788110DRS
10117-548EA 18105.92 CAD
10117-548
Legato 110 DRS Dual Rate Syringe Pump System, KD Scientific
Pumps Syringe Pumps
This syringe pump system allows the user to configure two different Legato 110 Infuse / Withdraw Programmable pumps with independent flow rates using the built in multistep programming and input / output signal communication
  • High resolution color touch screen, touch pad lock feature, LED light on front panel, built in RS285 interface, USB port
  • Full metal chassis, built in syringe table
  • Up to 30 lb. linear force
  • Advanced microstepping techniques
  • I/O and TTL interface, continuous mode of operation
  • Spill dam, programmable
All control is set through the innovative Legato touch screen user interface. No external computer is required. Accomodates syringe sizes 0.5ul to 60 ml. A communication cable for the two pumps is included.
KD Scientific syringe pumps are for laboratory use only. Not for clinical use.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.