VWR® Mechanical Cryogenic Freezer Boxes with Dividers
Supplier: VWR International
89214-766 89214-770 89214-772 89214-774
89214-766CS 483.33 CAD
89214-766 89214-770 89214-772 89214-774
VWR® Mechanical Cryogenic Freezer Boxes with Dividers
Boxes Cryoboxes
These Mechanical Cryogenic Freezer Boxes with dividers are constructed of durable fiberboard with a protective moisture repellent coating.
Two and three inch high boxes are available. VWR® Freezer Boxes can be used with most manufacturer's upright and chest freezer racks in general purpose and ultra low mechanical freezers.