Extension Support, For Heating Mantles, Ace Glass Incorporated
Supplier: Ace Glass
12095-04 12095-02 12095-12 12095-06 12095-08 12095-14 12095-10
89076-350EA 341.97 CAD
89076-350 89076-348 89076-346 89076-356 89076-344 89076-354 89076-352
Extension Support, For Heating Mantles, Ace Glass Incorporated
Supports Flask Supports
For heating mantles up to 5,000 mL capacity
Used on heating mantles with soft outer covering instead of spun aluminum shell.