Corning® Polypropylene Erlenmeyer Flasks, Corning
Supplier: Corning
4985P-50 4985P-100 4985P-250 4985P-500 4985P-1L
30617-086CS 109.16 CAD
30617-086 30617-088 30617-090 30617-092 30617-094
Corning® Polypropylene Erlenmeyer Flasks, Corning
Flasks Erlenmeyer Flasks
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
With blue, easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.  
Order Now
SPECIFICATIONS
Environmentally Preferable