Synthware Pressure Equalizing Funnel with Teflon® Metering Stopcock, Kemtech America
Supplier: Kemtech America
F611410 F611425 F611460 F612460 F614125 F614250 F614500
60004-290EA 380.53 CAD
60004-290 60004-292 60004-296 60004-300 60004-306 60004-310 60004-312
Synthware Pressure Equalizing Funnel with Teflon® Metering Stopcock, Kemtech America
Funnels Separating Funnels
With Teflon® adjustable bore metering plug
  • Metering stopcock
  • Heavy wall borosilicate glass
  • Graduated
  • Standard ground glass sizes available
The metering plug completely solves the problems of sudden overflow or non-flow during addition of reagents.