Synthware PTFE Flask Length Stopper, Kemtech America
Supplier: Kemtech America
S120013 S120016 S120022 S120027 S120038
60004-874EA 39.25 CAD
60004-874 60004-876 60004-878 60004-880 60004-882
Synthware PTFE Flask Length Stopper, Kemtech America
Stoppers PE/LDPE/HDPE Stoppers
Stoppers features PTFE (Polytetrafluoroethylene) handle for easy griping.
  • Teflon® construction
  • Standard joint sizes available
Ideal for use in flasks.