Synthware Flasks for Solvent Transfer/Storage, Kemtech America
Supplier: Kemtech America
F484250 F484258 F481500 F484500 F484508 F484000 F484008
60003-216EA 171.44 CAD
60003-216 60003-218 60003-220 60003-222 60003-226 60003-228 60003-230
Synthware Flasks for Solvent Transfer/Storage, Kemtech America
Reaction Vessels Reaction Vessels, Flask Shape
Flasks are equipped with a high vacuum valve that features a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
  • High Vacuum Valve included
  • Various sizes available
  • 14/20 and 24/40 ground glass joint available
Heavy wall synthware straus flask is commonly used for storing dired and degassed solvents.