Synthware Graham Condensers, Kemtech America
Supplier: Kemtech America
C141120 C149200 C144200 C144250 C144300 C144400
60000-922EA 259.42 CAD
60000-922 60000-924 60000-926 60000-928 60000-930 60000-932
Synthware Graham Condensers, Kemtech America
Condensers
Graham condenser with 10 mm or 8 mm hose connection and ground glass joints.
  • available in 3 different standard ground glass joint sizes
  • Jacketed condensor body
  • Various sizes available
Synthware Graham condenser is designed to efficiently cool and condense a gas back into a liquid during the chemical distillation process.