Synthware Vacuum/Inert Gas Adapters, 90°, Kemtech America
Supplier: Kemtech America
A921420 A921922 A922440
60000-180EA 44.48 CAD
60000-180 60000-182 60000-184
Synthware Vacuum/Inert Gas Adapters, 90°, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Adapters for vacuum/inert gas applications.
  • 90° angle
  • Top hose connection
  • Bottom male [ST] joint
Vacuum/inert gas adapters feature a 90° angle hose connection on the top and a male joint.