VWR® Benchtop Chillers, 240V
Supplier: VWR International
97058-182 97058-186 97058-194 97058-202 97058-206
97058-182EA 8404.59 CAD
97058-182 97058-186 97058-194 97058-202 97058-206
VWR® Benchtop Chillers, 240V
Circulating Thermostats Closed Circulator Systems Cooling Circulators
VWR® Benchtop Chillers are environmentally friendly and economical alternatives to tap-water cooling.
  • Maximize bench space while maintaining optimal cooling and pumping power
  • Easy-to-read LED display
  • Front-mounted air filter keeps chiller running at peak efficiency
The unique design of VWR® Benchtop Chillers maximizes precious bench space without compromising cooling and pumping power.
Features include large easy-to-read LED displays, space-saving designs, and cooling at ambient temperatures as high as 35 °C.
The top-mounted fill port with spill protection cup prevents damage from potential spills when filling. The front-mounted easy-access air filter keeps the chiller running at peak efficiency.
Certifications: CE marked. cTUVus compliant.
Ordering information: Choice of pumps (magnetic drive centrifugal, turbine) and compressor sizes (1/2, 1/3, 1/10 HP). Chillers are supplied with two 1.3 cm (1/2") NPT x 1.3 cm (1/2") HB tubing adapters, two 1.3 cm (1/2") NPT x 9.5 cm (3/8") HB tubing adapters, and power cord. Prior to ordering, please check the specific plug configuration needed for your electrical requirements. European plug on 240V models. Other country-specific plugs are also available.
Caution: Note these are all 240 V, 50 Hz units, the corresponding 120 V units are 97058-180, 97058-184, 97058-192, 97058-200 and 97058-204 respectively.