Shandon™ Bluing Reagent, Epredia
Supplier: RICHARD-ALLAN SCIENTIFIC CA
6769001
6769002
CA89369-344CS
251.95
CAD
CA89369-344
CA89369-346
Shandon™ Bluing Reagent, Epredia
Stains
Microscopy and staining reagents
Employ a gentle, pH-controlled solution for bluing hematoxylin with the Epredia™ Shandon™ Bluing Reagent.
- Ready-to-use
- Thoroughly pH-controlled for completely effective bluing
It replaces the strong alkaline rinses or long tap water washes which may cause loss of cells or tissue sections.