168-Place Cupellation Furnace, Furnace and Assay Supplies
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
FAS-180-OIL FAS-180-NG1 FAS-180-LPG1 FAS-180-NG2 FAS-180-LPG2
CA11023-220EA 125233.53 CAD
CA11023-220 CA11023-222 CA11023-224 CA76047-120 CA76047-122
168-Place Cupellation Furnace, Furnace and Assay Supplies
Ovens Furnaces (Ashing and Muffle Ovens)
These 168-place cupellation furnaces are available with LPG, natural gas, or diesel fuel source options. Cupels should be 168 x 6A, 7A, or 7AS. Optional port viewing window, rear extraction unit and hand valve door control extras are also available.
  • Suitable for large labs 400-1200 samples per day including standards
  • Base construction uses high-quality 100 x 100 RHS
  • Furnace body construction is durable 5mm laser-cut plate
  • High quality refractory brick and insulation
  • Bolt holes in base plates of all 4 legs for individual adjustment on uneven surfaces
More Product Information