25/42 Place Fusion Furnaces, Furnace and Assay Supplies
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
FAS-110-NG1 FAS-110-OIL FAS-110-LPG1 FAS-110-NG2 FAS-110-LPG2
CA11023-238EA 105603.11 CAD
CA11023-238 CA11023-240 CA11023-242 CA76047-128 CA76047-130
25/42 Place Fusion Furnaces, Furnace and Assay Supplies
Ovens Furnaces (Ashing and Muffle Ovens)
These 25/42-place fusion furnaces are available with LPG, natural gas or diesel fuel source options. Crucibles should be 25 x 50g or 42 x 40g.
  • Suitable for small labs; 100-400 samples per day including standards
  • Base construction uses high-quality 75 x 75 RHS
  • Furnace body construction is durable 5mm laser-cut plate
  • High quality refractory brick and insulation
  • Bolt holes in base plates of all 4 legs for individual adjustment on uneven surfaces
  • Pneumatic door controls with foot pedal controller—available with an optional hand valve controller
This furnace has a fan forced single burner system with an automatic igniton system
The door has a pneumatic ram opening system with an easy to use foot pedal control or optional hand valve that allows you to open the door part-way so that you can check inside with minimal temperature loss.
Temperature is monitored and modulated automatically by a Type K thermocouple system and Omron temperature controller pre-set for a maximum temperature of 1200 degrees Celsius. This controller is simple to operate, and has a 2-level display (PV and SV) that shows actual temperature as well as the temperature the furnace is set to.
Ordering information: For optional equipment, contact your sales representative.
Caution: Please note that a minimum of 1 meter clearance around the furnace is required.