Silica sand 30 Mesh

Supplier: Avantor
Danger

CA10153-802EA 157.35 CAD
CA10153-802 80705-016
Silica sand 30 Mesh
Silica

Silica is an acidic component found in fire assay flux.


In either sand or fine powder format, it is added to samples to protect the crucible in the fusion phase of the Fire Assay process. Insufficient silica levels will result in increased and erratic crucible wear.

Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1427 °C
Density: 2.622…2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 112945-52-5
EINECS: 238-878-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR