Potassium nitrate (nitre- KNO₃)

Supplier: Avantor

Synonyms: Nitrate of Potash

AMAS029-942-01
80705-020EA 323.95 CAD
80705-020
Potassium nitrate (nitre- KNO₃)
Potassium nitrate
Formula: KNO₃
MW: 101.1 g/mol
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
MDL Number: MFCD00011409
CAS Number: 7757-79-1
UN: 1486
ADR: 5.1,III
Merck Index: 13,07733

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR