Alpha Refrigerated Circulators, Lauda-Brinkmann
Supplier: Lauda-Brinkmann
L000650 L000651 L000652
97039-888EA 10354.59 CAD
97039-888 97039-890 97039-892
Alpha Refrigerated Circulators, Lauda-Brinkmann
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The Lauda-Brinkmann Alpha heating and cooling circulators provide a temperature range from -25 to 100 °C, depending on the exact model. This temperature range was selected with a focus of addressing most of the common temperature control applications within the laboratory. This entry level equipment range is made possible by minimizing any unnecessary features, with the focus on reliability and user-friendliness.
The Alpha heating and cooling circulators are suitable for operation with non-flammable liquids (water, water/glycol) and for both internal and external temperatureMore Product Information