Alpha Heating Circulators, Lauda-Brinkmann
Supplier: Lauda-Brinkmann
L000631 L000633
97039-882EA 5378.26 CAD
97039-882 97039-884 97039-886
Alpha Heating Circulators, Lauda-Brinkmann
Circulating Thermostats Closed Circulator Systems Heating Circulators
The LAUDA Alpha heating and cooling circulators provide a temperature range from -25 to 100 °C, depending on the exact model
The Alpha heating and cooling circulators are suitable for operation with non-flammable liquids (water, water/glycol) and for both internal and external temperature control tasks. A one point calibration of the thermostats can be carried out by the user. Stainless steel bath.
This temperature range was selected with a focus of addressing most of the common temperature control applications within the laboratory.   This entry level equipment range is made possible by minimizing any unnecessary features, with the focus on reliability and user-friendliness.