Tacky Traxx Mats, Tackified Cleanroom Entrance Mats, High-Tech Conversions
Supplier: High-Tech Conversions
TT-1836-34W TT-1836-34B TT-2436-34W TT-2436-34B TT-2645-34W TT-2645-34B TT-3636-34W TT-3636-34B TT-3645-34W TT-3645-34B TT-3660-34W TT-3660-34B TT-3636-34G TT-3645-34G TT-2436-34G TT-2445-34G TT-2545-34G TT-2645-34G TT-2645-64G TT-1836-34G TT-1845-34G TT-1845-34B TT-2445-34B TT-3645-64B TT-2445-34W
10127-156CS 305.29 CAD
10127-156 10127-158 10127-160 10127-162 10127-164 10127-166 10127-168 10127-170 10127-172 10127-174 10127-176 10127-178 10799-708 10799-710 10799-716 10863-324 10863-326 10863-328 10863-330 75815-618 76200-336 76200-338 76200-340 76292-876 76292-878
Tacky Traxx Mats, Tackified Cleanroom Entrance Mats, High-Tech Conversions
Floor Mats Tacky Mats
These mats effectively captures dirt and dust from foot traffic and equipment before it enters controlled environment.
  • Multi-layered, clean-film, full adhesive layers
  • Each layer peels off easily to reveal a new clean surface
  • Numbered corner tabs make tracking remaining sheets easy
  • Smooth, effective surface is not damaged by shoes, booties or cart wheels
  • Peel-off sheets eliminate the need for messy cleaning and make it easy to maintain a clean surface
  • Perfect for cleanrooms, laboratories and any other dust or dirt free environments