BRAND BRANDplates® inertGrade™ Microplates for Cell Culture, Sterile, BrandTech®
Supplier: Brandtech
781911 781910 781900 781902
10124-666PK 2827.93 CAD
10124-666 10124-736 10124-744 10124-746
BRAND BRANDplates® inertGrade™ Microplates for Cell Culture, Sterile, BrandTech®
Microplates Cell Culture Plates
BRANDplates® inertGrade™ plates are sterile plates treated for the cultivation of non-adherent cell lines and suspension cultures
  • For non-adherent and suspension cultures
  • Minimizes cell adhesion and protein absorbtion
  • Inhibits early differentiation of stem cells
  • Sterilized with ethylene oxide
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. The 96-well plates have an orange embossed alphanumeric code for easy well identification.
The surface is optimized to reduce cell adhesion, protein adsorption, enzyme and cellular activation, making this an ideal microplate surface for tumor and stem cell research. All plates are made of virgin polystyrene and sterilized by ethylene oxide.
Ordering information: All plates are individually wrapped with lid in packs of 40.