Thermal-Fused Laminate Lockers with Integrated Base, Wisconsin Bench
Supplier: Wisconsin Bench (WB)
1TFL-156018/C 2TFL-156018/C 1TFL-157218/C 5TFL-157218/C 2TFL-157218/C 3TFL-157218/C
10054-760EA 4523.46 CAD
10054-760 10054-762 10054-764 10054-950 10055-002 10055-004
Thermal-Fused Laminate Lockers with Integrated Base, Wisconsin Bench
Furniture Lockers
Impact resistant thermal-fused laminate (TFL) lockers features a 0.125" reveal, nickel finish Blum European style clip hinge with 120 degree opening, adjustable in all planes of operation and right mount typical.
  • Lockers available in tier one, two, three and five door configurations
  • Locker sides, backs and partitions are constructed using 45 lbs. density industrial grade particle board core
  • Two hinges are installed on doors up to 42"H, three hinges on doors that exceed 42"H
  • Integrated 4" base with levelers in each corner which easily adjust from the inside
More Product Information