Hardwood Bench Kits with Painted Metal Pedestals, 9.5" Width, Wisconsin Bench
Supplier: Wisconsin Bench (WB)
LBSMP9H048 LBSMP9H060 LBSMP9H072 LBSMP9H096
10054-654KT 2307.46 CAD
10054-654 10054-656 10054-658 10054-660
Hardwood Bench Kits with Painted Metal Pedestals, 9.5" Width, Wisconsin Bench
Furniture Gowning Benches
1.25" thick face-glued hardwood locker room bench with 2 or 3 almond painted steel 16.25" H pedestals.
  • Eased edges and corners for comfort and durability
  • Pedestals are 14-gauge steel with 8.25" plate top and bottom for support
9.5" W locker bench is finished with clear catalyzed lacquer two coats top and one coat bottom.
Ordering information: 9.5W×1.25" thick bench with 2 almond pedestals for lengths of 48", 60" and 72" and 3 almond pedestals for 96" L. Hardware to secure bench to pedestal are included and should be installed with no more than 4 feet span between pedestals. Hardware to mount kit to the floor are not included. Ships in two cartons. Dimensions given are as installed.