Ergonomic Shovel, Small Blade, Remco
Supplier: Remco
56115
76008-476EA 137.43 CAD
76008-476
Ergonomic Shovel, Small Blade, Remco
Spades and Shovels
This robust ergonomic shovel is designed for use in direct contact with food.
The handle is reinforced with an inner aluminum tube. It is stepped in two places which helps to prevent back injuries. The neck joint has been strengthened in order to withstand heavy duty use. The long and small blade combination makes it suitable for shoveling smaller volumes and reaching into deep containers.
Certifications: FDA compliant raw material.