VWR® Petri Dishes, Nesting Alpine
Supplier: VWR
3060-00360
CA73370-042CS 309.11 CAD
CA73370-042
VWR® Petri Dishes, Nesting Alpine
Culture Dishes Petri Dishes
The unique recessed lid on these dishes offers excellent stacking stability and saves space.
  • Recessed lid design
  • Excellent stackability
  • Compact design
  • Flat bottom
They work with most models of media pouring equipment
The flat bottom allows a reduction in the amount of media required. Patented cross ribs on the bottom make these dishes resilient to warpage when media is poured at a high temperature. These dishes are excellent for performing colony counts because the cross ribs also provide a useful border for making quadrents.
Available with four ventilation plugs on the underside of the lid allow for venting and prevent condensation from developing and dripping onto the culture.