Batch Ball and Rod Mills, Sepor
Supplier: SEPOR CA
010E-007 010E-008 010E-013 010E-014
CA10118-260EA 4801.64 CAD
CA10118-260 CA10118-262 CA10118-264 CA10118-266
Batch Ball and Rod Mills, Sepor
Mills
Designed to be companion equipment to the SEPOR drive rolls, these durable containers will grind from 1000 to 2500 g charges.
  • In the stainless steel models, all wetted parts are stainless steel
  • Grinding media is also available
Heavy-duty construction, including machined rolling rings, a quick-threaded removable yoke, a rubber gasketed lid with handles, and interior lifter bars. Both ball and rod mills are available in mild steel, 1/4" urethane-lined mild steel, and stainless steel constructions.
Accessories information: The charge for ball mills isMore Product Information