Cupel Fork, Webber's Welding
Supplier: Webber's Welding
107654-07 FA116A FA116B FA116S
CA11017-646EA 435.34 CAD
CA11017-646 CA11017-650 CA11017-652 CA11019-812
Cupel Fork, Webber's Welding
Forks
Cupel Fork is specially designed for heavy-duty applications against heat from ovens or furnaces, while its mild steel prongs provide durable and dependable service.
Ordering information: Overall length: 57"
Handle length: 46"