Hollow Cathode Lamps to Suit PerkinElmer Systems, Photron
Supplier: PHOTRON CA
P914 P921 P926 P933 P951
CA11018-216EA 1042.33 CAD
CA11018-216 CA11018-230 CA11018-240 CA11018-254 CA11018-290
Hollow Cathode Lamps to Suit PerkinElmer Systems, Photron
Lights Hollow Cathode Lamps
Photron's P900 (51mm or 2.0” Diameter) series is designed for direct use in AAS systems manufactured by PerkinElmer.
These hollow cathode lamps are produced to provide direct use without adapters in all PerkinElmer AAS instruments. All windows on these lamps are fully fused and contain no "gassy" adhesives which drastically reduce shelf life. Totally hermetic glass seals ensures a clean fill gas and cathode for the life of the lamp and most importantly a shelf life longer than 5 years. The application of the same glass cathode shielding technique used in all Photron hollow cathode lampsMore Product Information