Tetrahydrofuran ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
89703-700
89703-540
CA71009-096EA
1430.94
CAD
CA71009-096
CA11021-160
Tetrahydrofuran ACS
Tetrahydrofuran
Formula:
C₄H₈O Boiling Pt: 66 °C (1013 hPa) Melting Pt: –108.5 °C Density: 0.888 g/cm³ (20 °C) Flash Pt: –21.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005356 CAS Number: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3,II REACH: 05-2114615188-48 Merck Index: 12,09351 |
|
Specification Test Results
Assay (C4H8O) | 99.0% min. |
Colour (APHA) | 20 max. |
Peroxide (as H2O2) | 0.015% max. |
Residue after ignition | 0.03% max. |
Water (H2O) | 0.05% max. |
Stabilized with BHT | Confirmed |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...