VWR® Traceable® Refrigerator/Freezer Digital Thermometer
Supplier: VWR International
89094-778 61161-364
89094-778EA 221.05 CAD
89094-778 61161-364
VWR® Traceable® Refrigerator/Freezer Digital Thermometer
Thermometers Digital Thermometers
Accurately monitor temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with this enclosed temperature-buffered sensor.
  • Probe is sealed in glycol solution
  • Min/max memory
  • High and low settable alarms
Bottle insulates sensor from rapid temperature changes when refrigerator door is opened.
Triple display simultaneously shows high, low, and current temperatures. Min/max monitors high/low readings overnight, on weekends, or for any time period—a significant advantage over current reading-only glass thermometers. Range is -58 to 158°F and -50 to 70°C with a resolution of 1° and accuracy of ±1°C.
Alarm feature provides alert when temperature rises above or falls below a set point. Alarm is programmable in 1° increments. Visual and audible signals continue even if temperature returns to non-alarm range. Ten-foot micro-cable permits refrigerator doors to close on it.
Solid-state probe eliminates mercury contamination in refrigerators. Probe is sealed in a miniature bottle (1 x 2-1/2 inches) filled with nontoxic glycol. Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration). Eliminates concerns about contact with food or drinking water.
Certifications: To assure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO/IEC 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Technology).