Cell Strainers
Supplier: Corning
431751 431752 431750
10054-456CS 176.41 CAD
10054-456 10054-458 10054-462
Cell Strainers
Cell Strainers
Corning® cell strainers are sterile, easy to use devices for rapidly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Fits 50 ml Falcon® conical tubes.
  • Strong nylon mesh for optimal performance in a variety of applications
  • Extended lip on strainer enables aseptic handling with forceps
  • Moulded color-coded polypropylene frame with tab enables easy handling and identification
  • Sterilised by gamma irradiation, non cytotoxic
  • Conveniently accessible in individual packaging
Use these versatile cell strainers toMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable