Puritan® Small Cotton Tipped Applicator, Puritan Medical Products
Supplier: Puritan Medical Products
868-PCS 868-PCS 868-WCS
89237-580CS 903.71 CAD
89237-580 CA82030-716 82030-716
Puritan® Small Cotton Tipped Applicator, Puritan Medical Products
Swabs Swabs for Cleaning
This cotton tipped applicator is produced with High Quality USP grade cotton which is carded and coiled onsite to maintain consistency
Small cotton tip ideal for hard-to-reach or small areas. Applications to include general purpose swab, cleaning medical devices, removing dirt or oil from machine parts, veterinary care, and cleaning sensors and components.
It is bonded securely to a sturdy wood handle by an aqueous based adhesive.
Certifications: CE marked.
Ordering information: Made in the USA.