Puritan® Opti-Swab® Liquid Amies Collection and Transport System, Puritan Medical Products
Supplier: Puritan Medical Products
LA-106 LA-116 LA-117
89239-114CS 701.96 CAD
89239-114 89239-116 89239-118 CA89239-114
Puritan® Opti-Swab® Liquid Amies Collection and Transport System, Puritan Medical Products
Swabs Transport Swabs
Opti-Swab® liquid amies collection and transport systems are uniquely engineeded to provide superior performance and ease of use in the collection and transport of clinically significant bacteria.
Togethe, proprietary patented HydraFlock® swabs combined with modified liquid amies tranport medium provide an ideal envrionment to protect, aerobic, anaerobic, and fastidious bacterial viability during specimen collection and transport.
These systems are offered with or without a swab. The vial has a leak-proof, self-centering cap.
Certifications: CE marked.
Ordering information: Products are final sale.