VWR® Magnifier Lamp
Supplier: VWR
3358
10048-662EA 269.92 CAD
10048-662
VWR® Magnifier Lamp
Magnifiers
Magnifier Lamp is the ideal tool to view objects at two times magnification in bright light, with high contrast and a sharply defined focus.
  • Bright light
  • Large viewing area
  • Vertical and horizontal clamps
  • High contrast and sharply defined focus
Designed specifically for lab and plant applications requiring precise work
Large viewing area (4 5/8") is easy on the eyes. Highly polished optical glass 3X (3-power) lens provides a crisp and distinct image with vivid enhancement of surface details. Lower energy consumption and long-life 9.4 watt LED bulb light the exact area to be viewed. Extension: 31 inches. Clamps vertically or horizontally to the top or side of any edge.
Viewing area measures 4 5/8" in diameter. Supplied with a 9.4 watt LED bright bulb.
Useful in inspection, electronics, quality control, sample examination, and biological analysis. Clamps vertically or horizontally. Control knobs permit locking in place. Designed for lab and plant applications.
Ordering information: Supplied with a clamp to affix lamp to any bench or surface.