Allophycocyanin (Crosslinked APC), Columbia Biosciences
Supplier: Columbia Biosciences
D3-000
D3-010
10065-578EA
143.15
CAD
10065-578
10065-580
Allophycocyanin (Crosslinked APC), Columbia Biosciences
Protein Linking and Labeling Systems
Allophycocyanin (APC) is a commonly used label in Time-Resolved Fluorescence Resonant Energy Transfer (TR-FRET) assays for High Throughput Screening and Flow Cytometry.